TCIAH0570

H0570-25ML
TCH0570-025MLEA 80.15 USD
TCH0570-025ML
TCIAH0570
CAS Number: 13074-65-2
MDL Number: MFCD00059516
Molecular Formula: C11H20O
Molecular Weight: 168.28
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 246
Flash Point (°C): 112
Specific Gravity (20/20): 0.89
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR