2-Heptylcyclopentanone ≥99.0%

Supplier: TCI America
Warning

H0569-25ML H0569-500ML
TCH0569-025MLEA 403 USD
TCH0569-025ML TCH0569-500ML
2-Heptylcyclopentanone ≥99.0%
2-Heptylcyclopentanone

CAS Number: 137-03-1
MDL Number: MFCD00019315
Molecular Formula: C12H22O
Molecular Weight: 182.31
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 264
Specific Gravity (20/20): 0.88

Formula: C₁₂H₂₂O
MW: 182.31 g/mol
Boiling Pt: 264 °C
Density: 0.88 (20 °C)
MDL Number: MFCD00019315
CAS Number: 137-03-1
EINECS: 205-273-1
https://us.vwr.com/stibo/web/std.lang.all/05/86/28970586.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR