1-Hexadecyne ≥90.0%

Supplier: TCI America

H0433-5ML
TCH0433-005MLEA 117.03 USD
TCH0433-005ML
1-Hexadecyne ≥90.0%
1-Hexadecyne

CAS Number: 629-74-3
MDL Number: MFCD00015084
Molecular Formula: C16H30
Molecular Weight: 222.42
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 284
Flash Point (°C): 108
Freezing point (°C): 13
Specific Gravity (20/20): 0.80


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Tetradecylacetylene

Formula: C₁₆H₃₀
MW: 222.41 g/mol
Boiling Pt: 284 °C
Melting Pt: 15 °C
Density: 0.80 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00015084
CAS Number: 629-74-3
EINECS: 211-106-3

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR