2-Heptyne ≥99.0%

Supplier: TCI America

H0431-5ML
TCH0431-005MLEA 120.18 USD
TCH0431-005ML
2-Heptyne ≥99.0%
2-Heptyne

CAS Number: 1119-65-9
MDL Number: MFCD00039970
Molecular Formula: C7H12
Molecular Weight: 96.17
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 11
Specific Gravity (20/20): 0.75


Other Product Names / Synonyms: n-Butylmethylacetylene

Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 110 °C
Melting Pt: 1 °C
Density: 0.75 (20 °C)
Flash Pt: 11 °C
MDL Number: MFCD00039970
CAS Number: 1119-65-9
EINECS: 214-285-6
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR