2-Hydroxy-3-methoxy-5-nitrobenzaldehyde ≥99.0% (by GC, titration analysis)

Supplier: TCI America

H0397-5G
TCH0397-005GEA 77.04 USD
TCH0397-005G
2-Hydroxy-3-methoxy-5-nitrobenzaldehyde ≥99.0% (by GC, titration analysis)
2-Hydroxy-3-methoxy-5-nitrobenzaldehyde

CAS Number: 17028-61-4
MDL Number: MFCD00017033
Molecular Formula: C8H7NO5
Molecular Weight: 197.15
Purity/Analysis Method: 99.0% (GC,T)
Form: Crystal
Melting point (°C): 140


Other Product Names / Synonyms: 2-Hydroxy-3-methoxy-5-nitrobenzaldehyde, 3-Methoxy-5-nitrosalicylaldehyde, 5-Nitro-o-vanillin

Formula: C₈H₇NO₅
Melting Pt: 140 °C
MDL Number: MFCD00017033
CAS Number: 17028-61-4
https://us.vwr.com/stibo/web/std.lang.all/04/19/28970419.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR