Hydroxyacetone ≥80.0%

Supplier: TCI America

H0388-100ML H0388-500ML H0388-25ML
TCH0388-500MLEA 224.07 USD
TCH0388-500ML TCH0388-100ML TCH0388-25ML
Hydroxyacetone ≥80.0%
Hydroxyacetone

CAS Number: 116-09-6
MDL Number: MFCD00004669
Molecular Formula: C3H6O2
Molecular Weight: 74.08
Purity/Analysis Method: 80.0% (GC)
Form: Clear Liquid
Flash Point (°C): 57
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Acetol, Hydroxy-2-propanone

Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –17 °C
Density: 1.08 (20 °C)
Flash Pt: 57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004669
CAS Number: 116-09-6
EINECS: 204-124-8
UN: 1224
ADR: 3,III
Merck Index: 13,00066
https://us.vwr.com/stibo/web/std.lang.all/04/14/28970414.jpg

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR