2'-Hydroxychalcone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2'-Hydroxybenzylidenacetophenone 1-(2-Hydroxyphenyl)-3-phenyl-2-propenone

H0385-25G
TCH0385-025GEA 72.05 USD
TCH0385-025G
2'-Hydroxychalcone ≥98.0% (by HPLC)
2'-Hydroxychalcone

CAS Number: 1214-47-7
MDL Number: MFCD00016441
Molecular Formula: C15H12O2
Molecular Weight: 224.26
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 88


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals, Bioscience, Aromatic Polyketides
Other Product Names / Synonyms: 2-Benzal-2'-hydroxyacetophenone, 2-Benzylidene-2'-hydroxyacetophenone

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Melting Pt: 88 °C
MDL Number: MFCD00016441
CAS Number: 1214-47-7
EINECS: 214-928-0
https://us.vwr.com/stibo/web/std.lang.all/04/12/28970412.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR