n-Heneicosane ≥99.0%

Supplier: TCI America

H0367-25G H0367-1G
TCH0367-025GEA 261.26 USD
TCH0367-025G TCH0367-001G
n-Heneicosane ≥99.0%
n-Heneicosane

CAS Number: 629-94-7
MDL Number: MFCD00009346
Molecular Formula: C21H44
Molecular Weight: 296.58
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 356
Melting point (°C): 41
Flash Point (°C): 166


Other Product Names / Synonyms: Henicosane

Formula: H₃C(CH₂)₁₉CH₃
MW: 296.58 g/mol
Boiling Pt: 356 °C
Melting Pt: 41 °C
Density: 0.7919 g/cm³ (20 °C)
Flash Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00009346
CAS Number: 629-94-7
EINECS: 211-118-9
https://us.vwr.com/stibo/web/std.lang.all/03/99/28970399.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR