2-Hydroxyacetophenone ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Hydroxyacetophenoneα-HydroxyacetophenonePhenacyl alcoholBenzoylcarbinol2-Hydroxy-1-phenylethanonePhenacyl hydroxyide

H0347-25G H0347-5G
TCH0347-25GEA 204.02 USD
TCH0347-25G TCH0347-005G
2-Hydroxyacetophenone ≥98.0%
2-Hydroxyacetophenone

CAS Number: 582-24-1
MDL Number: MFCD00041829
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 126
Melting point (°C): 88


Other Product Names / Synonyms: Benzoylcarbinol, Phenacyl Alcohol

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 126 °C
Melting Pt: 88 °C
Density: 1.0963 g/cm³ (99 °C)
MDL Number: MFCD00041829
CAS Number: 582-24-1
EINECS: 209-480-8
https://us.vwr.com/stibo/web/std.lang.all/03/86/28970386.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR