4-Pyridone ≥99.0%

Supplier: TCI America
Warning

Synonyms: 1,4-Dihydro-4-PyridinonePyridin-4-olγ-Pyridone4-Hydroxypyridine4-Pyridinol

H0332-25G
TCH0332-025GEA 81.2 USD
TCH0332-025G
4-Pyridone ≥99.0%
4-Pyridone

CAS Number: 626-64-2
MDL Number: MFCD00040458
Molecular Formula: C5H5NO
Molecular Weight: 95.10
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 235
Melting point (°C): 148
Flash Point (°C): 221


Other Product Names / Synonyms: 4-Pyridinol, 4-Pyridone

Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 235 °C
Melting Pt: 148 °C
Flash Pt: 221 °C
MDL Number: MFCD00006419
CAS Number: 626-64-2
EINECS: 210-958-3
https://us.vwr.com/stibo/web/std.lang.all/03/73/28970373.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR