8-Quinolinol ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
8-Hydroxyquinoline
CAS Number: 148-24-3
MDL Number: MFCD00006807
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 126
Melting point (°C): 75
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes, Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: 8-Hydroxyquinoline, Oxine, 8-Quinolinone, Quinophenol
Formula:
C₉H₇NO MW: 145.16 g/mol Boiling Pt: 126 °C Melting Pt: 75 °C Density: 1.03 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006807 CAS Number: 148-24-3 EINECS: 205-711-1 Merck Index: 13,04869 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...