4-Methyl-2-quinolinol ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 4-Methylquinolin-2-ol2-Lepidinol2-Hydroxy-4-methylquinoline

H0259-25G
TCH0259-025GEA 97.41 USD
TCH0259-025G
4-Methyl-2-quinolinol ≥98.0% (by HPLC, total nitrogen)
4-Methyl-2-quinolinol

CAS Number: 607-66-9
MDL Number: MFCD00006745
Molecular Formula: C10H9NO
Molecular Weight: 159.19
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 227


Other Product Names / Synonyms: 2-Hydroxylepidine

Formula: C₁₀H₉NO
MW: 159.185 g/mol
Boiling Pt: 346 °C (1013 hPa)
Melting Pt: 227 °C
MDL Number: MFCD00006745
CAS Number: 607-66-9
EINECS: 210-139-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR