4-Hydroxycoumarin ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Hydroxy-2-benzopyrone
H0235-25G
H0235-250G
TCH0235-250GEA
269.99
USD
TCH0235-250G
TCH0235-025G
4-Hydroxycoumarin ≥98.0% (by GC, titration analysis)
4-Hydroxycoumarin
CAS Number: 1076-38-6
MDL Number: MFCD00006856
Molecular Formula: C9H6O3
Molecular Weight: 162.14
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 212
Formula:
C₉H₆O₃ MW: 162.14 g/mol Boiling Pt: 349 °C (1013 hPa) Melting Pt: 212 °C |
MDL Number:
MFCD00006856 CAS Number: 1076-38-6 EINECS: 214-060-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...