1-Hexanethiol ≥96.0%

Supplier: TCI America

Synonyms: 1-Mercaptohexane n-Hexanethiol n-Hexyl mercaptan

H0103-25ML H0103-500ML
TCH0103-500MLEA 253.67 USD
TCH0103-500ML TCH0103-025ML
1-Hexanethiol ≥96.0%
1-Hexanethiol

CAS Number: 111-31-9
MDL Number: MFCD00004909
Molecular Formula: C6H14S
Molecular Weight: 118.24
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Melting point (°C): -81
Flash Point (°C): 30
Specific Gravity (20/20): 0.84


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Hexanethiol

Formula: C₆H₁₄S
MW: 118.24 g/mol
Boiling Pt: 150 °C
Melting Pt: –81 °C
Density: 0.84 (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00004909
CAS Number: 111-31-9
EINECS: 203-857-0
UN: 3071
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/01/65/28970165.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR