Hexamethylbenzene ≥99.0%

Supplier: TCI America

H0087-5G
TCH0087-005GEA 82.57 USD
TCH0087-005G
Hexamethylbenzene ≥99.0%
Hexamethylbenzene

CAS Number: 87-85-4
MDL Number: MFCD00008523
Molecular Formula: C12H18
Molecular Weight: 162.28
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 165

Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 165 °C
Density: 1.063 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008523
CAS Number: 87-85-4
EINECS: 201-777-0
https://us.vwr.com/stibo/web/std.lang.all/01/50/28970150.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR