Perchloroethane ≥99.0%

Supplier: TCI America

Synonyms: Hexachloroethane

H0060-100G H0060-25G H0060-500G
TCH0060-100GEA 30.22 USD
TCH0060-100G TCH0060-500G TCH0060-25G
Perchloroethane ≥99.0%
Perchloroethane

CAS Number: 67-72-1
MDL Number: MFCD00000799
Molecular Formula: C2Cl6
Molecular Weight: 236.72
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 185


Other Product Names / Synonyms: Carbon Hexachloride

Formula: C₂Cl₆
MW: 236.74 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 185 °C
Density: 2.091 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000799
CAS Number: 67-72-1
EINECS: 200-666-4
Merck Index: 13,04698
https://us.vwr.com/stibo/web/std.lang.all/01/29/28970129.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR