1-Heptanethiol ≥98.0%

Supplier: TCI America

Synonyms: 1-Mercaptoheptane n-Heptanethiol n-Heptyl mercaptan

H0029-10G H0029-25G
TCH0029-010GEA 91.35 USD
TCH0029-010G TCH0029-025G
1-Heptanethiol ≥98.0%
1-Heptanethiol

CAS Number: 1639-09-4
MDL Number: MFCD00004911
Molecular Formula: C7H16S
Molecular Weight: 132.27
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Melting point (°C): -43
Flash Point (°C): 46
Specific Gravity (20/20): 0.84


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Heptyl Mercaptan

Formula: C₇H₁₆S
MW: 132.27 g/mol
Boiling Pt: 177 °C
Melting Pt: –43 °C
Density: 0.84 (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00004911
CAS Number: 1639-09-4
EINECS: 216-678-8
UN: 3071
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/01/08/28970108.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR