Glutarimide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

G0269-5G
TCG0269-005GEA 42.68 USD
TCG0269-005G
Glutarimide ≥98.0% (by total nitrogen basis)
Glutarimide

CAS Number: 1121-89-7
MDL Number: MFCD00006670
Molecular Formula: C5H7NO2
Molecular Weight: 113.12
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 155


Other Product Names / Synonyms: 2,6-Diketopiperidine, 2,6-Piperidinedione

Formula: C₅H₇NO₂
Melting Pt: 155 °C
MDL Number: MFCD00006670
CAS Number: 1121-89-7
EINECS: 214-340-4
https://us.vwr.com/stibo/web/std.lang.all/98/97/28969897.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR