7-Isopropyl-1,4-dimethylazulene ≥99.0%

Supplier: TCI America
Warning

Synonyms: Guaiazulene

G0228-10G
TCG0228-010GEA 71.03 USD
TCG0228-010G
7-Isopropyl-1,4-dimethylazulene ≥99.0%
7-Isopropyl-1,4-dimethylazulene

CAS Number: 489-84-9
MDL Number: MFCD00003811
Molecular Formula: C15H18
Molecular Weight: 198.31
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 320
Flash Point (°C): 113
Freezing point (°C): 29


Product Categories: Bioscience, Sesquiterpenes
Other Product Names / Synonyms: 1,4-Dimethyl-7-isopropylazulene

Formula: C₁₅H₁₈
MW: 198.31 g/mol
Boiling Pt: 320 °C
Melting Pt: 133 °C
Density: 0.9759 g/cm³ (18 °C)
Flash Pt: 113 °C
MDL Number: MFCD00003811
CAS Number: 489-84-9
EINECS: 207-701-2
Merck Index: 12,04581
https://us.vwr.com/stibo/web/std.lang.all/98/65/28969865.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR