Guanidinium carbonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Guanidine carbonateGuanidine carbonate saltDiguanidinium carbonate

G0161-25G G0161-500G
TCG0161-500GEA 38.59 USD
TCG0161-500G TCG0161-025G
Guanidinium carbonate ≥98.0% (by titrimetric analysis)
Guanidinium carbonate

CAS Number: 593-85-1
MDL Number: MFCD00013029
Molecular Formula: CH5N3
Molecular Weight: 180.17
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 333

Formula: C₃H₁₂N₆O₃
MW: 180.17 g/mol
Melting Pt: 333 °C
Density: 1.251 g/cm³ (20 °C)
MDL Number: MFCD00013029
CAS Number: 593-85-1
EINECS: 209-813-7
https://us.vwr.com/stibo/web/std.lang.all/98/20/28969820.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR