Guaifenesin ≥98.0% (by HPLC and UV method)
Supplier: TCI America
Synonyms:
3-(2-Methoxyphenoxy)-1,2-propanediol
, 3-(2-Methoxyphenoxy)propane-1,2-diol
, Guaiacol glyceryl ether
G0159-25G
G0159-500G
TCG0159-500GEA
154.22
USD
TCG0159-500G
TCG0159-025G
Guaifenesin ≥98.0% (by HPLC and UV method)
Guaifenesin
CAS Number: 93-14-1
MDL Number: MFCD00016873
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 215
Melting point (°C): 83
Other Product Names / Synonyms: Glyceryl Guaiacolate, 3-(o-Methoxyphenoxy)-1,2-propanediol
Formula:
C₁₀H₁₄O₄ MW: 198.22 g/mol Boiling Pt: 215 °C Melting Pt: 83 °C |
MDL Number:
MFCD00016873 CAS Number: 93-14-1 EINECS: 202-222-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...