TCIAF0248

F0248-5G
TCF0248-005GEA 72.86 USD
TCF0248-005G
TCIAF0248
CAS Number: 451-69-4
MDL Number: MFCD00004370
Molecular Formula: C9H7FO2
Molecular Weight: 166.15
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 180
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR