Piperidine-N-carbaldehyde ≥98.0%

Supplier: TCI America

Synonyms: N-Formylpiperidine1-Formylpiperidine

F0234-25ML F0234-100ML F0234-500ML
TCF0234-25MLEA 25.68 USD
TCF0234-25ML TCF0234-500ML TCF0234-100ML
Piperidine-N-carbaldehyde ≥98.0%
Piperidine-N-carbaldehyde

CAS Number: 2591-86-8
MDL Number: MFCD00006483
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 222
Flash Point (°C): 91
Specific Gravity (20/20): 1.02


Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: 1-Piperidinecarboxaldehyde

Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 222 °C
Density: 1.02 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00006483
CAS Number: 2591-86-8
EINECS: 219-986-0
https://us.vwr.com/stibo/web/std.lang.all/88/37/28968837.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR