3-Fluoro-L-tyrosine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0201-100MG
TCF0201-100MGEA 138.6 USD
TCF0201-100MG
3-Fluoro-L-tyrosine ≥98.0% (by HPLC, titration analysis)
3-Fluoro-L-tyrosine

CAS Number: 7423-96-3
MDL Number: MFCD00002607
Molecular Formula: C9H10FNO3
Molecular Weight: 199.18
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -9 deg (C=1, 1mol/L HCl)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Tyr(3-F)-OH

Formula: C₉H₁₀FNO₃
MDL Number: MFCD00002607
CAS Number: 7423-96-3
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR