2-Fluorobenzothiazole ≥99.0%

F0180-1G
TCF0180-001GEA 102.12 USD
TCF0180-001G
2-Fluorobenzothiazole ≥99.0%
2-Fluorobenzothiazole

CAS Number: 1123-98-4
MDL Number: MFCD00047254
Molecular Formula: C7H4FNS
Molecular Weight: 153.17
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 93
Specific Gravity (20/20): 1.33
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Benzothiazolyl Fluoride

Formula: C₇H₄FNS
MW: 153.18 g/mol
Boiling Pt: 98 °C
Density: 1.33 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00047254
CAS Number: 1123-98-4
EINECS: 214-384-4
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/87/88/28968788.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR