4-Fluoro-3-nitroaniline ≥98.0%
Supplier: TCI America
|
Warning
|
F0168-25G
F0168-10G
TCF0168-010GEA
25.53
USD
TCF0168-010G
TCF0168-025G
4-Fluoro-3-nitroaniline ≥98.0%
4-Fluoro-3-nitroaniline
CAS Number: 364-76-1
MDL Number: MFCD00007833
Molecular Formula: C6H5FN2O2
Molecular Weight: 156.12
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 97
Flash Point (°C): 91
Formula:
C₆H₅FN₂O₂ MW: 156.12 g/mol Melting Pt: 97 °C Density: 0.43 g/cm³ (20 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007833 CAS Number: 364-76-1 EINECS: 206-665-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...