3-Fluorobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

F0158-25G F0158-250G
TCF0158-025GEA 35.53 USD
TCF0158-025G TCF0158-250G
3-Fluorobenzoic acid ≥98.0% (by titrimetric analysis)
3-Fluorobenzoic acid

CAS Number: 455-38-9
MDL Number: MFCD00002489
Molecular Formula: C7H5FO2
Molecular Weight: 140.11
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 123

Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00002489
CAS Number: 455-38-9
EINECS: 207-248-0
https://us.vwr.com/stibo/web/std.lang.all/87/73/28968773.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR