4-Fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
F0112-25G
TCF0112-025GEA
47.71
USD
TCF0112-025G
4-Fluorobenzoic acid ≥98.0% (by GC, titration analysis)
4-Fluorobenzoic acid
CAS Number: 456-22-4
MDL Number: MFCD00002530
Molecular Formula: C7H5FO2
Molecular Weight: 140.11
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 186
Formula:
C₇H₅FO₂ MW: 140.11 g/mol Boiling Pt: 254 °C (1013 hPa) Melting Pt: 186 °C Density: 1.49 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002530 CAS Number: 456-22-4 EINECS: 207-259-0 Merck Index: 13,04198 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...