Ethyl-2-furoate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl furan-2-carboxylateEthyl2-furoate

F0098-25G
TCF0098-025GEA 25.68 USD
TCF0098-025G
Ethyl-2-furoate ≥98.0%
Ethyl 2-furoate

CAS Number: 614-99-3
MDL Number: MFCD00003237
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 196
Flash Point (°C): 70
Freezing point (°C): 34
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Ethyl 2-Furoate, 2-Furancarboxylic Acid Ethyl Ester, 2-Furoic Acid Ethyl Ester

Formula: C₇H₈O₃
MW: 140.14 g/mol
Boiling Pt: 196 °C
Flash Pt: 70 °C
MDL Number: MFCD00003237
CAS Number: 614-99-3
EINECS: 210-404-0
https://us.vwr.com/stibo/web/std.lang.all/87/26/28968726.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR