Methyl formate ≥98.0%

Supplier: TCI America
Danger

Synonyms: Formic acid methyl ester

F0086-100ML
TCF0086-100MLEA 53.59 USD
TCF0086-100ML
Methyl formate ≥98.0%
Methyl formate

[for Spectrophotometry]
CAS Number: 107-31-3
MDL Number: MFCD00003291
Molecular Formula: C2H4O2
Molecular Weight: 60.05
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 32
Flash Point (°C): -20
Specific Gravity (20/20): 0.97
Storage Temperature: 0-10°C


Product Categories: Analytical Science, Solvents for Spectrophotometry
Other Product Names / Synonyms: Formic Acid Methyl Ester

Formula: C₂H₄O₂
MW: 60.05 g/mol
Boiling Pt: 32 °C
Melting Pt: –100.4 °C
Density: 0.97 (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003291
CAS Number: 107-31-3
EINECS: 203-481-7
UN: 1243
ADR: 3,I
Merck Index: 12,06153
https://us.vwr.com/stibo/web/std.lang.all/87/17/28968717.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR