Ethyl formate ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Formic acid ethyl ester
F0085-100ML
TCF0085-100MLEA
100.49
USD
TCF0085-100ML
Ethyl formate ≥98.0%
Ethyl formate
[for Spectrophotometry]
CAS Number: 109-94-4
MDL Number: MFCD00003294
Molecular Formula: C3H6O2
Molecular Weight: 74.08
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 54
Melting point (°C): -80
Flash Point (°C): -20
Specific Gravity (20/20): 0.92
Product Categories: Analytical Science, Solvents for Spectrophotometry
Other Product Names / Synonyms: Formic Acid Ethyl Ester
Formula:
C₃H₆O₂ MW: 74.08 g/mol Boiling Pt: 54 °C Melting Pt: –80 °C Density: 0.92 (20 °C) Flash Pt: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003294 CAS Number: 109-94-4 EINECS: 203-721-0 UN: 1190 ADR: 3,II Merck Index: 13,03841 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...