Methyl-2-furoate ≥99.0%

Supplier: TCI America

Synonyms: Methyl furan-2-carboxylateMethyl 2-Furancarboxylate

F0075-250ML F0075-25ML
TCF0075-250MLEA 159.31 USD
TCF0075-250ML TCF0075-025ML
Methyl-2-furoate ≥99.0%
Methyl 2-furoate

CAS Number: 611-13-2
MDL Number: MFCD00003236
Molecular Formula: C6H6O3
Molecular Weight: 126.11
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Flash Point (°C): 73
Specific Gravity (20/20): 1.18


Other Product Names / Synonyms: 2-Furoic Acid Methyl Ester, 2-Furancarboxylic Acid Methyl Ester, Methyl 2-Furoate

Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 181 °C
Density: 1.18 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00003236
CAS Number: 611-13-2
EINECS: 210-254-6
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/87/07/28968707.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR