Diethyl fumarate ≥98.0%

Supplier: TCI America
Warning

Synonyms: Fumaric acid diethyl ester

F0068-25ML F0068-500ML
TCF0068-500MLEA 89.32 USD
TCF0068-500ML TCF0068-025ML
Diethyl fumarate ≥98.0%
Diethyl fumarate

CAS Number: 623-91-6
MDL Number: MFCD00064455
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 219
Melting point (°C): 2
Flash Point (°C): 111
Specific Gravity (20/20): 1.06


Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Fumaric Acid Diethyl Ester

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 219 °C
Melting Pt: 2 °C
Density: 1.06 (20 °C)
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00064455
CAS Number: 623-91-6
EINECS: 210-819-7
https://us.vwr.com/stibo/web/std.lang.all/87/01/28968701.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR