Isopropyl formate ≥97.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Formic acid isopropyl ester
F0056-25ML
F0056-500ML
TCF0056-500MLEA
298.41
USD
TCF0056-500ML
TCF0056-25ML
Isopropyl formate ≥97.0%
Isopropyl formate
CAS Number: 625-55-8
MDL Number: MFCD00014127
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 68
Flash Point (°C): -6
Specific Gravity (20/20): 0.88
Other Product Names / Synonyms: Formic Acid Isopropyl Ester
Formula:
C₄H₈O₂ MW: 88.11 g/mol Boiling Pt: 68 °C Melting Pt: –89 °C Density: 0.88 (20 °C) Flash Pt: –6 °C |
MDL Number:
MFCD00014127 CAS Number: 625-55-8 EINECS: 210-901-2 UN: 1281 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...