Butyl formate ≥95.0%

Supplier: TCI America
Danger

Synonyms: Formic acid butyl ester

F0051-500ML F0051-25ML
TCF0051-500MLEA 206.05 USD
TCF0051-500ML TCF0051-25ML
Butyl formate ≥95.0%
Butyl formate

CAS Number: 592-84-7
MDL Number: MFCD00003296
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Melting point (°C): -91
Flash Point (°C): 16
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: Formic Acid Butyl Ester

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 107 °C
Melting Pt: –91 °C
Density: 0.90 (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00003296
CAS Number: 592-84-7
EINECS: 209-772-5
UN: 1128
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/86/89/28968689.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR