2-Fluorotoluene ≥99.0%

Supplier: TCI America

Synonyms: 1-Fluoro-2-methylbenzene

F0039-500ML F0039-25ML
TCF0039-25MLEA 16.24 USD
TCF0039-25ML TCF0039-500ML
2-Fluorotoluene ≥99.0%
2-Fluorotoluene

CAS Number: 95-52-3
MDL Number: MFCD00000322
Molecular Formula: C7H7F
Molecular Weight: 110.13
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 114
Flash Point (°C): 12
Specific Gravity (20/20): 1.00

Formula: C₇H₇F
MW: 110.13 g/mol
Boiling Pt: 114 °C
Melting Pt: –62 °C
Density: 1.00 (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00000322
CAS Number: 95-52-3
EINECS: 202-428-5
UN: 2388
ADR: 3,II
Merck Index: 13,04207
https://us.vwr.com/stibo/web/std.lang.all/86/80/28968680.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR