4-Fluoroaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

F0033-25ML
TCF0033-25MLEA 50.75 USD
TCF0033-25ML
4-Fluoroaniline ≥98.0% (by GC, titration analysis)
4-Fluoroaniline

CAS Number: 371-40-4
MDL Number: MFCD00007829
Molecular Formula: C6H6FN
Molecular Weight: 111.12
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 188
Melting point (°C): -1
Flash Point (°C): 73
Specific Gravity (20/20): 1.16

Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 188 °C
Melting Pt: –1 °C
Density: 1.16 (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007829
CAS Number: 371-40-4
EINECS: 206-735-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,04196
https://us.vwr.com/stibo/web/std.lang.all/86/76/28968676.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR