Triacetoxyethylsilane ≥96.0%

Supplier: TCI America

Synonyms: Ethylsilanetriyl triacetateTriacetoxy(ethyl)silane

E0525-25G
TCE0525-25GEA 40.6 USD
TCE0525-25G
Triacetoxyethylsilane ≥96.0%
Triacetoxyethylsilane

CAS Number: 17689-77-9
MDL Number: MFCD00053703
Molecular Formula: C8H14O6Si
Molecular Weight: 234.28
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 109
Flash Point (°C): 106
Freezing point (°C): 8
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: Ethyltriacetoxysilane

Formula: C₈H₁₄O₆Si
MW: 234.28 g/mol
Boiling Pt: 109 °C
Density: 1.15 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD00053703
CAS Number: 17689-77-9
EINECS: 241-677-4
https://us.vwr.com/stibo/web/std.lang.all/80/34/28968034.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR