2-Ethyl-1,3-cyclopentanedione ≥97.0% (by GC, titration analysis)
Supplier: TCI America
E0507-25G
E0507-5G
TCE0507-005GEA
34
USD
TCE0507-005G
TCE0507-025G
2-Ethyl-1,3-cyclopentanedione ≥97.0% (by GC, titration analysis)
2-Ethyl-1,3-cyclopentanedione
CAS Number: 823-36-9
MDL Number: MFCD00044191
Molecular Formula: C7H10O2
Molecular Weight: 126.16
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 175
Formula:
C₇H₁₀O₂ MW: 126.16 g/mol Melting Pt: 175 °C |
MDL Number:
MFCD00044191 CAS Number: 823-36-9 EINECS: 212-512-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...