Ethylmagnesium bromide 13% ∼1 mol/L in THF

E0497-250G
TCE0497-250GEA 146.73 USD
TCE0497-250G
Ethylmagnesium bromide 13% ∼1 mol/L in THF
Ethylmagnesium bromide

CAS Number: 925-90-6
MDL Number: MFCD00000043
Molecular Formula: C2H5BrMg
Molecular Weight: 133.27
Flash Point (°C): -14
Specific Gravity (20/20): 1.01

Formula: C₂H₅BrMg
Density: 1.01 (20 °C)
Flash Pt: –14 °C
MDL Number: MFCD00000043
CAS Number: 925-90-6
EINECS: 213-127-3
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR