7-Ethoxy-4-methylcoumarin ≥97.0%

Supplier: TCI America

E0491-25G
TCE0491-025GEA 181.5 USD
TCE0491-025G
7-Ethoxy-4-methylcoumarin ≥97.0%
7-Ethoxy-4-methylcoumarin

CAS Number: 87-05-8
MDL Number: MFCD00016971
Molecular Formula: C12H12O3
Molecular Weight: 204.23
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 111

Formula: C₁₂H₁₂O₃
Melting Pt: 111 °C
MDL Number: MFCD00016971
CAS Number: 87-05-8
EINECS: 201-721-5
https://us.vwr.com/stibo/web/std.lang.all/80/05/28968005.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR