1,2-Epoxyhexane ≥96.0%
Supplier: TCI America
Synonyms:
Butyloxirane
E0445-25ML
TCE0445-25MLEA
63.12
USD
TCE0445-25ML
1,2-Epoxyhexane ≥96.0%
1,2-Epoxyhexane
CAS Number: 1436-34-6
MDL Number: MFCD00005154
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 15
Specific Gravity (20/20): 0.83
Other Product Names / Synonyms: 1,2-Hexylene Oxide
Formula:
C₆H₁₂O Boiling Pt: 120 °C Density: 0.83 (20 °C) Flash Pt: 15 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005154 CAS Number: 1436-34-6 EINECS: 215-864-6 UN: 1993 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...