4-Ethylanisole ≥98.0%
Supplier: TCI America
Synonyms:
1-Ethyl-4-methoxybenzene
E0436-25ML
TCE0436-025MLEA
88.31
USD
TCE0436-025ML
4-Ethylanisole ≥98.0%
4-Ethylanisole
CAS Number: 1515-95-3
MDL Number: MFCD00015176
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 79
Specific Gravity (20/20): 0.96
Other Product Names / Synonyms: 1-Ethyl-4-methoxybenzene
Formula:
C₉H₁₂O MW: 136.19 g/mol Boiling Pt: 195 °C Density: 0.96 (20 °C) Flash Pt: 79 °C |
MDL Number:
MFCD00015176 CAS Number: 1515-95-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...