Triethyl phosphonoformiate

Supplier: TCI America

Synonyms: Phosphonoformic acid, triethyl esterEthyl diethoxyphosphinylformateEthyl (diethoxyphosphoryl)formate

E0400-25G
TCE0400-025GEA 39 USD
TCE0400-025G
Triethyl phosphonoformiate
Triethyl phosphonoformiate

CAS Number: 1474-78-8
MDL Number: MFCD00009108
Molecular Formula: C7H15O5P
Molecular Weight: 210.17
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 133
Flash Point (°C): 119
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: Diethoxyphosphinylformic Acid Ethyl Ester, (Ethoxycarbonyl)phosphonic Acid Diethyl Ester, Triethyl Phosphonoformate

Formula: C₇H₁₅O₅P
MW: 210.166 g/mol
Boiling Pt: 133 °C
Density: 1.15 (20 °C)
Flash Pt: 119 °C
MDL Number: MFCD00009108
CAS Number: 1474-78-8
EINECS: 216-016-8
https://us.vwr.com/stibo/web/std.lang.all/79/20/28967920.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR