4-Ethylbenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

E0379-25G
TCE0379-025GEA 70 USD
TCE0379-025G
4-Ethylbenzoic acid ≥98.0% (by GC, titration analysis)
4-Ethylbenzoic acid

CAS Number: 619-64-7
MDL Number: MFCD00002570
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 113

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 113 °C
Density: ∼0.9961 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002570
CAS Number: 619-64-7
EINECS: 210-605-3
https://us.vwr.com/stibo/web/std.lang.all/79/02/28967902.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR