N-Ethyl-o-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(Ethylamino)tolueneN-Ethyl-2-methylaniline

E0177-500ML E0177-25ML
TCE0177-25MLEA 21.32 USD
TCE0177-25ML TCE0177-500ML
N-Ethyl-o-toluidine ≥98.0% (by GC, titration analysis)
N-Ethyl-o-toluidine

CAS Number: 94-68-8
MDL Number: MFCD00026695
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 98
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: N-Ethyl-2-methylaniline, 2-(Ethylamino)toluene

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 214 °C
Melting Pt: –15 °C
Density: 0.95 (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00026695
CAS Number: 94-68-8
EINECS: 202-354-3
UN: 2754
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/77/52/28967752.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR