4-Ethylpyridine ≥98.0%

Supplier: TCI America

Danger

E0170-500ML E0170-25ML
TCE0170-500MLEA 684.91 USD
TCE0170-500ML TCE0170-25ML
4-Ethylpyridine ≥98.0%
4-Ethylpyridine

CAS Number: 536-75-4
MDL Number: MFCD00006450
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 50
Specific Gravity (20/20): 0.94

Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 165 °C
Melting Pt: -91 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 50 °C
Storage Temperature: Ambient
CAS Number: 536-75-4
EINECS: 208-646-7
UN: 1993
ADR: 3,III
Merck Index: 13,03880
https://us.vwr.com/stibo/web/std.lang.all/77/45/28967745.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR