2-Ethylphenol ≥98.0%

Supplier: TCI America

Synonyms: 1-Ethyl-2-hydroxybenzene

E0160-100ML E0160-25ML E0160-500ML
TCE0160-100MLEA 108.57 USD
TCE0160-100ML TCE0160-025ML TCE0160-500ML
2-Ethylphenol ≥98.0%
2-Ethylphenol

CAS Number: 90-00-6
MDL Number: MFCD00002249
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 197
Melting point (°C): -18
Flash Point (°C): 78
Specific Gravity (20/20): 1.02

Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 197 °C
Melting Pt: –18 °C
Density: 1.02 (20 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00002249
CAS Number: 90-00-6
EINECS: 201-958-4
UN: 3145
ADR: 8,III
Merck Index: 13,03871
https://us.vwr.com/stibo/web/std.lang.all/77/38/28967738.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR