Ethylmagnesium bromide 39% ∼3 mol/L in ethyl ether

E0134-250G
TCE0134-250GEA 83.25 USD
TCE0134-250G
Ethylmagnesium bromide 39% ∼3 mol/L in ethyl ether
Ethylmagnesium bromide

CAS Number: 925-90-6
MDL Number: MFCD00000043
Molecular Formula: C2H5BrMg
Molecular Weight: 133.27
Flash Point (°C): -40

Formula: C₂H₅BrMg
Flash Pt: –40 °C
MDL Number: MFCD00000043
CAS Number: 925-90-6
EINECS: 213-127-3
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR