N-Ethylaniline ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Danger

E0059-25ML E0059-500ML
TCE0059-25MLEA 24.24 USD
TCE0059-25ML TCE0059-500ML
N-Ethylaniline ≥99.0% (by GC, titration analysis)
N-Ethylaniline

CAS Number: 103-69-5
MDL Number: MFCD00009025
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: 99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 204
Melting point (°C): -63
Flash Point (°C): 91
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: N-Ethyl-N-phenylamine

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 204 °C
Melting Pt: –63 °C
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
CAS Number: 103-69-5
EINECS: 203-135-5
UN: 2272
ADR: 6.1,III
Merck Index: 13,03800

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR